Learn More
KleenGuard™ Nemesis™ Eyewear
GREENER_CHOICE
KleenGuard™ Nemesis™ Safety Eyewear is stylish, durable and have a lightweight construction.
Supplier:  KleenGuard™ 25688
 View more versions of this product
Catalog No. 19-150-1659
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
KleenGuard™ Nemesis™ Safety Eyewear's stylish good looks and durable, lightweight construction are just some of the reasons why millions of people wear them for protection both at work and at home.
Nemesis™ Safety Glasses have polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss. The wraparound frame has a base curve of 8 that extends the range of peripheral protection, as well as soft touch temples for comfort.
The Nemesis™ safety glasses have a patented nose piece designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose – all for long-term comfort.
Each pair of KleenGuard™ Nemesis™ safety glasses come with a neck cord for securing them, so that the glasses are ready to protect at a moment's notice. The safety eyewear is manufactured to comply with the ANSI/ISEA Z87.1+ standard for personal eye protection.
Both men and women will find these stylish glasses to be comfortable and functional for construction, manufacturing, gun shooting / shooting range, and a host of other industries.
  • Ideal for indoor use
  • Feature a sporty, flexible, lightweight design that your employees will be happy to wear; they have a stylish, sporty look and feel
  • Meets ANSI Z87.1+ standards for personal eye protection
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC
  • Patented FLEX-DRY designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose – all for long-term comfort
Compliance
Meet ANSI/ISEA Z87.1+ standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UV-treated
Smoke Mirror
Black
Plastic
Plastic
Meets ANSI/ISEA Z87.1+ standards.
Safety Glasses
Neck Cord
Reusable
UV400
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.