Learn More
Ansell KleenGuard™ Nemesis™ Readers Safety Glasses
GREENER_CHOICE
Catalog No. 19005077 Shop All Ansell Products
Change view
Click to view available options
Magnification Power:
+1.50
+2.00
+2.50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
19-005-077 +1.50
19-005-078 +2.00
19-005-079 +2.50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-005-077 Supplier Ansell Supplier No. 22516
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard™ Nemesis™ Readers Safety Glasses feature durable lightweight construction for protection at work and at home.
  • Smoke lenses are ideal for outdoor use
  • Sporty, flexible, lightweight design that your employees will be happy to wear
  • Neck cord included
  • Meets ANSI Z87.1+ standards for personal eye protection
  • Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection and are hard coated for scratch resistance
  • FLEX-DRY™ nosepiece designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose – all for long-term comfort
  • Recyclable via The RightCycle Program
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Impact-resistant, UV-treated
Lens Type Magnifying
Certifications/Compliance ANSI Z87.1+
Lens Tint Smoke
For Use With (Application) Safety sunglasses with diopters
Frame Color Black
Magnification Power +1.50
No. per Case 12 pairs/Cs.
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.