Learn More
KleenGuard™ Nemesis™ Safety Glasses
GREENER_CHOICE
KleenGuard™ Nemesis™ Safety Glasses feature durable lightweight construction for protection at work and at home.
Supplier:  KleenGuard™ 22476
 View more versions of this product
Catalog No. 19-005-076
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Sporty, flexible, lightweight design
  • Optional Anti-Fog coating prevents fogging; ideal for humid environments and temperature variances
  • Meets ANSI Z87.1+ standards for personal eye protection
  • Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection
  • Patented FLEX-DRY™ nosepiece is designed to be flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
  • Recyclable via The RightCycle Program
  • Lens options include:
    • Smoke tint lenses with outer blue mirror finish coating is ideal for outdoor use where sunlight can cause eyestrain and fatigue
    • Copper blue shield lenses have a light brown tint and are designed to filter out blue light for enhanced sharpness and depth perception; ideal for low-light conditions
    • Bronze lenses provide visual definition with increased contrast and enhanced contours in most light conditions
    • Amber/yellow lenses block out blue light, making the environment appear brighter; ideal for low-light conditions and indoor use
    • IRUV Shade 3 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB) during tasks like light cutting and brazing
    • IRUV Shade 5 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB) during tasks like medium cutting brazing
    • Brown polarized lenses are effective for filtering scattered blue light to improve depth perception and contrast; ideal for outdoor use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anti-fog, Impact-resistant, UV-treated
Amber
General-purpose eye protection
12 Pairs/Cs.
Reusable
Plastic
ANSI Z87.1+
Safety Glasses
Black
Plastic
Polycarbonate
UV400
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.