Learn More
KleenGuard™ Nemesis™ Safety Glasses
GREENER_CHOICE
Catalog No. 19706110 Shop All Ansell Products
Change view
Click to view available options
Frame Color:
Black
Brown
Camouflage
Gun Metal
Red
Lens Tint:
Amber
Blue, Mirror, Smoke
Bronze
Brown
Clear
Shade 3
Shade 5
Smoke
Lens Coating:
Anti-fog, Impact-resistant, UV-treated
Impact-resistant, UV-treated
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-706-110 Camouflage Smoke Anti-fog, Impact-resistant, UV-treated
19-170-103 Black Amber Impact-resistant, UV-treated
19-005-063 Black Blue, Mirror, Smoke Impact-resistant, UV-treated
19-005-064 Black Brown Impact-resistant, UV-treated
19-005-076 Black Amber Anti-fog, Impact-resistant, UV-treated
19-005-082 Black Shade 5 Impact-resistant, UV-treated
19-706-100 Camouflage Clear Impact-resistant, UV-treated
19-005-065 Camouflage Bronze Impact-resistant, UV-treated
19-005-080 Camouflage Amber Anti-fog, Impact-resistant, UV-treated
19-005-085 Gun Metal Smoke Impact-resistant, UV-treated
19-005-081 Red Smoke Anti-fog, Impact-resistant, UV-treated
19-005-086 Brown Brown Impact-resistant, UV-treated
19-005-084 Black Shade 3 Impact-resistant, UV-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Catalog No. 19-706-110 Supplier KleenGuard™ Supplier No. 22609
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard™ Nemesis™ Safety Glasses feature durable lightweight construction for protection at work and at home.
  • Sporty, flexible, lightweight design
  • Optional Anti-Fog coating prevents fogging; ideal for humid environments and temperature variances
  • Meets ANSI Z87.1+ standards for personal eye protection
  • Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection
  • Patented FLEX-DRY™ nosepiece is designed to be flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
  • Recyclable via The RightCycle Program
  • Lens options include:
    • Smoke tint lenses with outer blue mirror finish coating is ideal for outdoor use where sunlight can cause eyestrain and fatigue
    • Copper blue shield lenses have a light brown tint and are designed to filter out blue light for enhanced sharpness and depth perception; ideal for low-light conditions
    • Bronze lenses provide visual definition with increased contrast and enhanced contours in most light conditions
    • Amber/yellow lenses block out blue light, making the environment appear brighter; ideal for low-light conditions and indoor use
    • IRUV Shade 3 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB) during tasks like light cutting and brazing
    • IRUV Shade 5 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB) during tasks like medium cutting brazing
    • Brown polarized lenses are effective for filtering scattered blue light to improve depth perception and contrast; ideal for outdoor use
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Impact-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+
Lens Tint Smoke
Product Type Safety Glasses
For Use With (Application) Safety sunglasses
Frame Color Camouflage
No. per Case 12 Pairs/Cs.
Material Plastic
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Wrap-around
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.