Learn More
Ansell KleenGuard™ Nemesis™ VL Safety Glasses
GREENER_CHOICE
Catalog No. 19005071 Shop All Ansell Products
Change view
Click to view available options
For Use With (Application):
Cutting and brazing
General-purpose eye protection
Safety sunglasses
Frame Color:
Camouflage
Gun Metal
Lens Tint:
Blue, Mirror
Bronze, Indoor/Outdoor
IR/UV 5.0, Shade 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Frame Color Lens Tint
19-005-071 Safety sunglasses Gun Metal Blue, Mirror
19-005-073 Cutting and brazing Gun Metal IR/UV 5.0, Shade 5
19-005-072 General-purpose eye protection Camouflage Bronze, Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-005-071 Supplier Ansell Supplier No. 20471
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard™ Nemesis™ VL Safety Glasses feature durable lightweight construction for protection at work and at home.
  • Includes neck cord to keep glasses safe when not being worn
  • Extra-lightweight frameless design
  • No-brow design increases upward peripheral vision and limits interference with hard hat suspension systems
  • Meets ANSI Z87.1+ standards for personal eye protection
  • TAA compliant
  • 8.25 base curve polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection
  • Lightweight and frameless for long-term comfort
  • Recyclable via The RightCycle Program
  • Several lens options including:
    • Bronze lenses provide visual definition with increased contrast
    • IRUV Shade 5 lenses protect against infrared (IR) and ultraviolet radiation (UVA/UVB)
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Impact-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+
Lens Tint Blue, Mirror
For Use With (Application) Safety sunglasses
Frame Color Gun Metal
No. per Case 12 pairs/Cs.
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.