Learn More
Ansell KleenGuard™ V60 Dual Readers Safety Eyewear
When you need vision assistance and eye protection, you can trust KleenGuard Dual Readers. They feature integrated diopters (+1.5 strength) on both the top and the bottom for vision help when looking both up and down.
Supplier: Ansell 20387
This item is not returnable.
View return policy
Description
- Ideal for when workers need vision assistance and eye protection
- Stylish, nylon frames feature a soft, self-adjusting rubber nose pad for comfort
- Clear lenses allow maximum visible light transmission (ideal for indoor use) and are hard coated for scratch resistance and longer life
- Meet ANSI Z87.1+ standards for high impact
- Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
- Come in three different strengths (+1.5, +2.0 and +2.5)
- 7 base curve, which helps provide for peripheral vision and protection and feature an hard coating to prevent lens damage
Specifications
ANSI Z87.1 | |
Dual Readers Safety Eyewear | |
Black | |
UV400 |
Clear | |
For performing tasks that require up and down near vision or working upside down or overhead | |
+1.50 |