Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klenow Exo, 1000 U, G-Biosciences
SCI_ED

Catalog No. S001106
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
S001106 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001106 Supplier G-Biosciences Supplier No. 7862270
Add to Cart
Add to Cart

Klenow fragment (Exo-) is a mesophilic DNA polymerase deficient in both proofreading (3′→5′) and nick-translation (5′→3′) nuclease activities, with a moderate strand displacement activity during DNA synthesis.

  • The protein is expressed as a truncated product of the E. coli PolA gene and contains the D355A and E357A mutations to abolish 3'→5' exonuclease activity.

Specifications

Packaging Vial
Enzyme Exonuclease
Sensitive to Heat Inactivation Yes
Product Type Klenow Exo
Research Category Next-generation Sequencing, DNA Repair, Traditional Cloning
Product Line Thermo Scientific
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.