Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Klenow Fragment, LC (2 U/μL)

Catalog No. FEREP0054 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
300 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREP0054 300 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREP0054 Supplier Thermo Scientific™ Supplier No. EP0054
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Klenow Fragment is the Large Fragment of DNA Polymerase exhibiting 5'→3' polymerase, 3'→5' exonuclease (proofreading) activities, but lacking 5'→3' exonuclease activity.

Thermo Scientific Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5'→3' polymerase activity and 3'→5' exonuclease (proofreading) activity, but lacks 5'→3' exonuclease activity of DNA polymerase I.

Highlights

  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl-, biotin-, digoxigenin- and fluorescently-labeled nucleotides)
  • Active in restriction enzyme, PCR, RT, and T4 DNA Ligase buffers

Applications

  • DNA blunting by fill-in 3'-overhangs
  • Random-primed DNA labeling
  • Labeling by fill-in 5'-overhangs of dsDNA
  • DNA sequencing by the Sanger method
  • Site-specific mutagenesis of DNA with synthetic oligonucleotides
  • Second strand synthesis of cDNA
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Polymerase DNA Polymerase I
Quantity 300 units
Product Type Klenow Fragment
What are the differences between Thermo Scientific DNA Polymerase I, Klenow Fragment, and Klenow Fragment (exo-)?

Thermo Scientific DNA Polymerase I possesses 5′?3′ DNA synthesis activity, 3′?5′ exonuclease (proofreading) activity, and 5′?3′ exonuclease activity. Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5′?3′ polymerase activity and 3′?5′ exonuclease (proofreading) activity, but lacks 5′?3′ exonuclease activity. Klenow Fragment, exo-, is also the large fragment of DNA polymerase I. It exhibits 5′?3′ polymerase activity, but lacks the 3′?5′ and 5′?3′ exonuclease activities.

What conditions are optimal for fill-in reactions using Klenow (Large fragment of DNA Polymerase)?

Klenow can be used to "fill in" 5' overhangs of double-stranded DNA fragments using common restriction endonuclease buffers. We recommend REact 2 buffer (1X concentration is 50 mM Tris-HCl pH 8, 50 mM NaCl, 10 mM MgCl2) , but other buffers will also work.

Fill-in Reaction Conditions:

1. Dilute Large Fragment of DNA Polymerase I to 0.5 U/µL with Klenow Dilution Buffer.
2. To a 1.5-mL microcentrifuge tube on ice, add: 10X REact 2 Buffer - 3 µL, 0.5 mM dATP - 1 µL, 0.5 mM dCTP - 1 µL, 0.5 mM dGTP - 1 µL, 0.5 mM dTTP - 1 µL, DNA 0.5-1 µg, Large fragment of DNA Polymerase I - 1 µL, Autoclaved distilled water to 30 µL.
3. Mix gently and centrifuge briefly to bring the contents to the bottom of the tube.
4. Incubate at room temperature for 10-15 minutes or 20 minutes on ice.
5. Terminate fill-in reaction by phenol extraction.

To label the DNA fragment, use 1-2 µL of [alpha-32P]dNTP (400 Ci/mmol, 10 mCi/mL) (24-48 pmoles) instead of the corresponding cold dNTP.

Note: Thermo Fisher Scientific also offers an Exo minus Klenow, which is provided with its own buffers.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.