Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Klenow Fragment (10 U/μL)
GSA_VA

Catalog No. p-4521355
Change view
Click to view available options
Quantity:
1,500 units
300 units
Concentration:
10 U/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
FEREP0051 300 units 10 U/μL
FEREP0052 1,500 units 10 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEREP0051 Supplier Thermo Scientific™ Supplier No. EP0051
Only null left
Add to Cart
Edge
Add to Cart

Klenow Fragment is the Large Fragment of DNA Polymerase exhibiting 5'→3' polymerase, 3'→5' exonuclease (proofreading) activities, but lacking 5'→3' exonuclease activity.

Thermo Scientific Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5'→3' polymerase activity and 3'→5' exonuclease (proofreading) activity, but lacks 5'→3' exonuclease activity of DNA polymerase I.

Highlights

  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl-, biotin-, digoxigenin- and fluorescently-labeled nucleotides)
  • Active in restriction enzyme, PCR, RT, and T4 DNA Ligase buffers

Applications

  • DNA blunting by fill-in 3'-overhangs
  • Random-primed DNA labeling
  • Labeling by fill-in 5'-overhangs of dsDNA
  • DNA sequencing by the Sanger method
  • Site-specific mutagenesis of DNA with synthetic oligonucleotides
  • Second strand synthesis of cDNA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Polymerase DNA Polymerase I
Quantity 300 units
Product Type Klenow Fragment
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.