Learn More
Invitrogen™ KLF4 Monoclonal Antibody (1E5)

Description
MA5-15672 targets KLF4 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15672 immunogen is purified recombinant fragment of human KLF4 (Amino Acid: 2-470) expressed in E. Coli. MA5-15672 detects KLF4 which has a predicted molecular weight of approximately 55kDa.
Specifications
Specifications
| Antigen | KLF4 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 1E5 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | KLF4 |
| Gene Accession No. | O43474 |
| Gene Alias | endothelial Kruppel-like zinc finger protein; epithelial zinc finger protein EZF; Ezf; Gklf; gut Kruppel-like factor; gut-enriched krueppel-like factor; high mobility group nucleosomal binding domain 3; high mobility group nucleosome-binding domain-containing protein 3; HMGN3; KLF4; Krueppel-like factor 4; Kruppel like factor 4; Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); PNAS-24; PNAS-25; RP11-150J11.1; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP7; Zie |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.