Learn More
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (89%), Rat (89%), Cat (93%), Pig (93%), Sheep (92%), Rhesus Monkey (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | KLF4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | KLF4 |
| Gene Accession No. | O43474, Q60793 |
| Gene Alias | endothelial Kruppel-like zinc finger protein; epithelial zinc finger protein EZF; Ezf; Gklf; gut Kruppel-like factor; gut-enriched krueppel-like factor; high mobility group nucleosomal binding domain 3; high mobility group nucleosome-binding domain-containing protein 3; HMGN3; KLF4; Krueppel-like factor 4; Kruppel like factor 4; Kruppel-like factor 4 (Gut enriched kruppel-like factor) (Epithelial zinc-finger protein EZF); Kruppel-like factor 4 (gut); PNAS-24; PNAS-25; RP11-150J11.1; thyroid hormone receptor interacting protein 7; Thyroid receptor-interacting protein 7; TR-interacting protein 7; TRIP7; Zie |
| Gene Symbols | KLF4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
