Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF5 Recombinant Rabbit Monoclonal Antibody (23H10L5)

Catalog No. PI701885 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI701885 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI701885 Supplier Invitrogen™ Supplier No. 701885
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Monkey, Rabbit and Pig Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

KLF5 is a member of the Kruppel-like factor subfamily of zinc finger proteins. It contains three C2H2-type zinc fingers at the carboxyl-terminus that preferentially bind to cis-DNA elements that are GC rich. KLF5 is a transcription activator that promotes cell proliferation and oncogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF5
Applications ChIP Assay, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23H10L5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene KLF5
Gene Accession No. Q13887
Gene Alias (intestinal Kruppel-like factor; 4930520J07Rik; basic transcription element binding protein 2; basic transcription element binding protein BTEB2; basic transcription element-binding protein 2; BTEB2; BTE-binding protein 2; CKLF; colon krueppel-like factor; colon kruppel-like factor; GC box binding protein 2; GC-box-binding protein 2; IKLF; Intestinal-enriched krueppel-like factor; intestinal-enriched kruppel-like factor; Klf5; Klf5C isoform; Krueppel-like factor 5; Kruppel like factor 5; Kruppel-like factor 5; Kruppel-like factor 5 (intestinal); RP11-505F3.5; transcription factor BTEB2
Gene Symbols KLF5
Host Species Rabbit
Immunogen Peptides corresponding to Human KLF5 (aa 11-24, 358-372).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.