Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF6 Monoclonal Antibody (2A2), Active Motif™
SDP

Catalog No. p-7188779
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3314 10 μg
50-199-3313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3314 Supplier Active Motif Supplier No. 61298
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a member of the Kruppel-like family of transcription factors. The zinc finger protein is a transcriptional activator, and functions as a tumor suppressor. Multiple transcript variants encoding different isoforms have been found for this gene, some of which are implicated in carcinogenesis.

Specifications

Antigen KLF6
Applications Western Blot
Classification Monoclonal
Clone 2A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene Klf6
Gene Accession No. Q99612
Gene Alias Aa1017; AI448727; BCD1; B-cell-derived protein 1; C86813; CBA1; COPEB; core promoter element binding protein; Core promoter element-binding protein; CPBP; FM2; FM6; GBF; GC-rich binding factor; GC-rich sites-binding factor GBF; Ierepo1; Ierepo3; immediate early response, erythropoietin 1; KLF6; Krueppel-like factor 6; Kruppel like factor 6; Kruppel-like factor 6; Kruppel-like zinc finger protein Zf9; PAC1; Proto-oncogene BCD1; protooncogene B-cell derived 1; R75280; ST12; suppression of tumorigenicity 12 (prostate); Suppressor of tumorigenicity 12 protein; Transcription factor Zf9; ZF9
Gene Symbols Klf6
Host Species Mouse
Immunogen This kLF6 antibody was raised against a recombinant protein corresponding to amino acids 28-199 of human kruppel-like factor 6 (kLF6).
Purification Method Protein A
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1316
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.