Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF9 Monoclonal Antibody (OTI6A9), TrueMAB™, OriGene
SDP

Catalog No. 501682139
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2139 Supplier OriGene Supplier No. TA808446
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KLF-9 is a transcription factor belonging to the Sp 1 C2H2 type zinc finger family. It binds to GC box elements located in the promoter. This binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription. KLF-9 serves as a key transcriptional regulator for uterine endometrial cell proliferation, adhesion, and differentiation both during pregcy and tumorigenesis.

Specifications

Antigen KLF9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6A9
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KLF9
Gene Accession No. Q13886
Gene Alias BTEB, BTEB1
Gene Symbols KLF9
Host Species Mouse
Immunogen Full length human recombit protein of human KLF9 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 687
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.