Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLF9 Recombinant Rabbit Monoclonal Antibody (5H16L7)

Catalog No. 701888 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-188-8 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-188-8 Supplier Invitrogen™ Supplier No. 701888
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Bovine, Pig and Monkey. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

KLF-9 is a transcription factor belonging to the Sp 1 C2H2 type zinc finger family. It binds to GC box elements located in the promoter. This binding of the encoded protein to a single GC box inhibits mRNA expression while binding to tandemly repeated GC box elements activates transcription. KLF-9 serves as a key transcriptional regulator for uterine endometrial cell proliferation, adhesion, and differentiation both during pregnancy and tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF9
Applications ChIP Assay, Western Blot
Classification Recombinant Monoclonal
Clone 5H16L7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene KLF9
Gene Accession No. O35739, Q01713, Q13886
Gene Alias 2310051E17Rik; AA589643; basic transcription element binding protein 1; basic transcription element-binding protein 1; BTEB; BTEB1; BTEB-1; BTE-binding protein 1; ENSMUSG00000055440; GC-box-binding protein 1; Gm9971; KLF9; Krueppel-like factor 9; Kruppel like factor 9; Kruppel-like factor 9; transcription factor BTEB1
Gene Symbols KLF9
Host Species Rabbit
Immunogen Peptides corresponding to Human KLF9 (aa 34-49, 85-98).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117560, 16601, 687
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.