Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHDC5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554292 Supplier Invitrogen™ Supplier No. PA554292
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CKPQFHLLGS PPQAPGDVSL KQRLREARMT GTPVLVALGD FLGGPLAPHP YQGEPPSMLR YEEMTERWFP LANNLPPDLV NVRGYG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex required for mitotic progression and cytokinesis. The BCR(KLHL42) E3 ubiquitin ligase complex mediates the ubiquitination and subsequent degradation of KATNA1. Involved in microtubule dynamics throughout mitosis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHDC5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KLHL42
Gene Accession No. Q9P2K6
Gene Alias A330079N23Rik; AI450355; C230080I20Rik; Ctb9; cullin-3-binding protein 9; kelch domain containing 5; kelch domain-containing protein 5; kelch like family member 42; kelch-like 42; kelch-like family member 42; kelch-like family, member 42; kelch-like protein 42; KIAA1340; Klhdc5; KLHL42; mKIAA1340
Gene Symbols KLHL42
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLHDC5. Recombinant protein control fragment (Product #RP-93574).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57542
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.