Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLHL21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531730 Supplier Invitrogen™ Supplier No. PA531730
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, Jurkat, Raji. Predicted reactivity: Mouse (95%), Rat (94%), Xenopus laevis (80%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex required for efficient chromosome alignment and cytokinesis. The BCR(KLHL21) E3 ubiquitin ligase complex regulates localization of the chromosomal passenger complex (CPC) from chromosomes to the spindle midzone in anaphase and mediates the ubiquitination of AURKB. Ubiquitination of AURKB by BCR(KLHL21) E3 ubiquitin ligase complex may not lead to its degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLHL21
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.84 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene KLHL21
Gene Accession No. Q9UJP4
Gene Alias 1810045K06Rik; D330008A20; kelch like family member 21; kelch-like 21; kelch-like family member 21; kelch-like protein 21; KIAA0469; Klhl21; MGC99635; mKIAA0469; RGD1305863
Gene Symbols KLHL21
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KLHL21. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9903
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.