Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Klotho-derived peptide 1
GREENER_CHOICE

Catalog No. 78301
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-301 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 78-301 Supplier Tocris Bioscience™ Supplier No. 7830/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

TGF-β receptor 2 (TβR2) binding peptide; disrupts the TGF-β/TβR2 interaction

Klotho-derived peptide 1 (KP1) is a peptide that binds to TGF-β receptor 2 (TβR2) (Kd = 1.4 μM). It inhibits TGF-β signaling by blocking TGF-β/TβR2 interaction. In mouse models of renal fibrosis, intravenous injection of KP1 results in its specific accumulation in the injured kidneys. It suppresses TGF-β signaling, repressing fibroblast activation and ameliorates kidney fibrosis in vivo.

Specifications

Gene ID (Entrez) 9365
Purity 95%
Content And Storage Store at -20°C
Gene Alias EC 3.2.1, EC 3.2.1.31, KL, klotho, TGFB2, TGFbeta, TGFbeta 2
Molecular Weight (g/mol) M.W. (Observed): 3228.48
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.