Learn More
Cayman Chemical KlothodRIved PeptID 156 500ug
SDP
Supplier:  Cayman Chemical 38166500 UG
Encompass_Preferred
A metabolite of L-phenylalanine increases the proliferation of MCF-7 cells increases luteinizing hormone and estrogen levels and ERa and ERB expression in the uterus of immature female mice urinary levels are increased in male obese patients compared with non-obese males
Catalog No. 50-291-5612
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.