Learn More
Cayman Chemical ANTIBODY PF-9363 5mg
SDP
Supplier:  Cayman Chemical 377785 MG
Encompass_Preferred
A CK2 inhibitor (IC50 0.39 UM) selective for CK2 over PKA GSK3B LCK DYRK1A and PI3KY (IC50s 16 14 11 8 200) decreases the viability of Jurkat leukemia cells in a concentration-dependent manner
Catalog No. 50-291-5613
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.