Learn More
Invitrogen™ KLRG1 Monoclonal Antibody (2F1), Brilliant Violet™ 711, eBioscience™, Invitrogen™

Description
Specifications
Specifications
| Antigen | KLRG1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2F1 |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Violet 711 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Klrg1 |
| Gene Accession No. | O88713 |
| Gene Alias | 2F1; 2F1-Ag; CLEC15A; C-type lectin domain family 15 member A; C-type lectin domain family 15, member A; ITIM-containing receptor MAFA-L; killer cell lectin like receptor G1; killer cell lectin-like receptor subfamily G member 1; killer cell lectin-like receptor subfamily G, member 1; Klrg1; Mafa; MAFA-2F1; MAFAL; MAFA-L; MAFA-LIKE; MAFA-like receptor; Mast cell function-associated antigen; mast cell function-associated antigen (ITIM-containing); mast cell function-associated antigen 2F1; natural killer cell receptor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.