Learn More
TARGETMOL CHEMICALS INC KM02894 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7758125MG
Encompass_Preferred
Also available in 50 mg and bulk. Please contact Fisher for quotes. KM02894 is a glutamate release inhibitor. Cancer cells release high levels of glutamate that can disrupt normal bone turnover which can lead to cancer-induced bone pain.KM02894 may be used in the study of tumor-related diseases. purity: 98%
Catalog No. 50-257-9130
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.