Learn More
KN-62
Description
It is used as a potent and selective inhibitor of CaMKII. KN-62 has demonstrated potent and selective inhibition of Ca2+/calmodulin-dependent protein kinase II (CaMKII; Ki=0.9 ?M) which causes cells to arrest in the S phase. In HEK-293 cells expressing type III adenylyl cyclase (III-AC), 10 ?M of KN-62 blocked CaMKII phosphorylation of III-AC, and inhibited hormone-stimulated Ca2+ oscillations. Additionally, KN-62 is a potent non-competitive antagonist at the P2X7 receptor (IC50 = 15 nM).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a potent and selective inhibitor of CaMKII. KN-62 has demonstrated potent and selective inhibition of Ca2+/calmodulin-dependent protein kinase II (CaMKII; Ki=0.9 μM) which causes cells to arrest in the S phase. In HEK-293 cells expressing type III adenylyl cyclase (III-AC), 10 μM of KN-62 blocked CaMKII phosphorylation of III-AC, and inhibited hormone-stimulated Ca2+ oscillations. Additionally, KN-62 is a potent non-competitive antagonist at the P2X7 receptor (IC50 = 15 nM).
Solubility
Soluble in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin (0.93 mg/ml), methanol (10 mg/ml), DMSO (12 mg/ml), chloroform (20 mg/ml), ethanol (∼1 mg/ml), 1:2 DMF:PBS (pH 7.2) (∼0.3 mg/ml), and DMF & DMSO (∼30 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 65°C to 67°C |
| Color | Yellow |
| Linear Formula | NH2(CH2)10NH2 |
| Quantity | 10 mg |
| Solubility Information | Soluble in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin (0.93mg/ml),methanol (10mg/ml),DMSO (12mg/ml),chloroform (20mg/ml),ethanol (∽1mg/ml),1:2 DMF:PBS (pH 7.2) (∽0.3mg/ml),and DMF & DMSO (∽30mg/ml). |
| Formula Weight | 721.84 |
| Physical Form | Powder |
| Chemical Name or Material | KN-62 |
Safety and Handling
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.