Learn More
Apexbio Technology LLC KN-93 139298-40-1 25mg
Supplier: Apexbio Technology LLC A353225
KN-93 (CAS 139298-40-1) is a selective inhibitor of calcium/calmodulin-dependent protein kinase II (CaMKII) a serine/threonine kinase activated by calcium-bound calmodulin KN-93 inhibits CaMKII enzymatic activity with reported IC50 values of approximately 0 37 0 5 M in vitro and a Ki value of 2 58 M against cardiac CaMKII In NIH 3T3 cells KN-93 (12 M) induces G1 cell-cycle arrest and at higher concentrations (24 M) promotes apoptosis Due to CaMKII s role in neuronal function cardiac signaling immune activation and disease states including Alzheimer s and arrhythmias KN-93 serves as a tool compound for studying CaMKII-mediated processes in biomedical research
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.