Learn More
Apexbio Technology LLC KN-92 hydrochloride 1431698-47-3 50mg
SDP
Supplier:  Apexbio Technology LLC A353050
Encompass_Preferred
KN-92 hydrochloride is an inactive structural analog of the Ca /calmodulin-dependent protein kinase II (CaMKII) inhibitor KN-93 serving primarily as a comparative control compound in biological research Unlike KN-93 KN-92 hydrochloride lacks meaningful inhibitory activity against CaMKII exhibiting a much higher Ki value ( 100 M) In experimental models it is utilized to distinguish CaMKII-dependent molecular and physiological responses particularly in studies of early afterdepolarizations (EADs) and cardiac repolarization events Based on these properties KN-92 hydrochloride is applied as a negative control in cardiac electrophysiological research
Catalog No. 50-000-94275
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.