Learn More
Apexbio Technology LLC KN-92 phosphate 1135280-28-2 10mg
Supplier: Apexbio Technology LLC A353110
KN-92 phosphate is a small-molecule analog of the Ca2 /calmodulin-dependent kinase II (CaMKII) inhibitor KN-93 It is designed as an inactive control compound to distinguish CaMKII-specific effects in experimental settings KN-92 phosphate does not inhibit CaMKII activity and thus serves as a negative control for CaMKII inhibition studies Additionally KN-92 phosphate blocks certain voltage-gated potassium (Kv) channels such as Kv1 2 and Kv1 5 reducing steady-state current amplitudes upon depolarization Based on these pharmacological properties KN-92 phosphate holds research potential in investigating mechanisms underlying CaMKII-related signaling pathways and Kv channel functions
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.