Learn More
Apexbio Technology LLC Amsacrine 51264-14-3 25mg
SDP
Supplier:  Apexbio Technology LLC B130225
Encompass_Preferred
Amsacrine (CAS 51264-14-3) also known as m-AMSA is a small-molecule inhibitor of DNA topoisomerase II It is widely used as an antineoplastic agent in biomedical research particularly for investigating the treatment of lymphomas and acute leukemias Amsacrine exhibits cytotoxic effects across various cancer cell lines with reported IC50 values of 190 2 27 4 ng/ml 46 1 3 9 ng/ml and 22 6 3 1 ng/ml in HT1376 RT112 and RT4 bladder cancer cells and 11 8 2 0 ng/ml 5 0 0 4 ng/ml and 11 7 1 5 ng/ml in the 833K Susa and GH testicular cancer cell lines respectively Testicular cancer cell lines display greater sensitivity to Amsacrine-induced inhibition than bladder cancer cell lines This compound is valuable for studying DNA damage mechanisms and the cellular response to topoisomerase II inhibition
Catalog No. 50-000-92012
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.