Learn More
Abcam KN-93 Water Soluble, CaMK II Inhibitor, 1 mg
Description
- Molecular weight: 599 Da
- Purity: >99%
- Potent, specific and cell-permeable calmodulin-dependent protein kinase (CaMK) II inhibitor (IC50 = 370 nM)
- Potent and direct extracellular open channel voltage-gated potassium channel blocker (IC50 = 307 nM for Kv1.5; independent of CaM kinase II inhibition)
- Solubility: Water soluble
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | CaMK II, Kv1.5 |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Alternate Part Number | KN-93 |
| Formulation | 100% KN-93 (water soluble) |
| Quantity | 1 mg |
| For Use With (Application) | CaMK II inhibition, Potassium channel blocking |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.