Learn More
Cayman Chemical KN-93phosphate 10mg
SDP
Supplier:  Cayman Chemical 2147210 MG
Encompass_Preferred
A CaMKII inhibitor (Ki = 370 nM) that inhibits the α- and β-subunits of CaMKII
Catalog No. 50-298-7455