Learn More
BYD Care™ KN95 Respirator Mask
Breathable, comfortable and foldable with a head strap for a tight fit providing at least 95% filtration efficiency. This respirator is not NIOSH-approved.
Supplier: BYD Care™ DG3101
Description
Mask consists of three layers of high quality nonwoven materials: the outer and inner layers are made of polypropylene spunbond nonwoven fabric, and the middle layer is made of polypropylene melt-blown nonwoven fabric. This three layer construction provides at least 95% filtration efficiency against solid and liquid aerosols free of oil.
- Soft, skin-friendly, comfortable wearing experience and protection for daily usage
- Foldable design can cover nose and mouth with close fit to the face, providing respiratory protection to reduce exposure to certain airborne particles
- Elastic head straps are soft and gentle to skin, eliminating pressure behind the ears even when worn for a long time
- Adjustable built-in nose bridge
- Easy breathability
- Single-use level ll ear loop mask
Specifications
White | |
General Purpose Particulate Respirator | |
Yes | |
Non-sterile |
Polypropylene | |
95% | |
2 Pk. | |
Earloops with Fastener |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.