Learn More
Eisco™ Knife Switch
SCI_ED
Catalog No. S27758 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
S27758 1 Ea.
1 options
Catalog No. S27758 Supplier Eisco™ Supplier No. PH1029J
Add to Cart
Add to Cart
This Knife Switch demonstrates the concept of a single pole double throw knife switch.
  • Includes (3) 4 mm terminals for banana plugs
  • Base Dimensions: 3.75 x 2.75 x 1.38 in. (95.25 x 69.85 x 35 mm)
  • Power Source is required for operation, sold separately
  • CE Certification
Specifications
Product Type Knife Switch
Description CE Certification
For Use With (Application) Demonstrating The Concepts of a Single Pole, Double Throw Knife Switch
Includes (3) 4 mm Banana Plug Terminals
Quantity 1 Ea.