Learn More
Description
- High Lumens of 163
- Run Time on High of up to 3.75 hours
- Run Time on Low of up to 12.00 hours
- Beam Distance of 54 meters
- Industrial rechargeable Ni-Cd 4.8 volt battery
- IPX4 rated for water resistant operation
- Comes with a hand hook, microprocessor with controlled high- and low-intensity modes, an emergency flash mode, and a moonlight mode
Specifications
Specifications
| Weight (English) | 18.3 oz. |
| Weight (Metric) | 518.8 g |
| Color | Yellow |
| Description | Intrinsically safe |
| For Use With (Application) | Hazardous Environments |
| For Use With (Equipment) | Safety Equipment |
| Height (Metric) | 23.4 cm |
| Material | Nylon |
| Width (Metric) | 7.2 cm |
| Product Type | Flashlight |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.