Learn More
ABclonal Technology KO Validated NRF2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A357720UL
Encompass_Preferred
This gene encodes a transcription factor which is a member of a small family of basic leucine zipper (bZIP) proteins The encoded transcription factor regulates genes which contain antioxidant response elements (ARE) in their promoters many of these genes encode proteins involved in response to injury and inflammation which includes the production of free radicals Multiple transcript variants encoding different isoforms have been characterized for this gene
Catalog No. 50-000-13626
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.