Learn More
Koehler™ Instrument LPG Copper Strip Corrosion Test Cylinder and Accessories
$61.10 - $2794.80
Specifications
Certifications/Compliance | ASTM D1838, GPA 2140, ISO 6251, CE |
---|---|
Material | Stainless Steel |
Catalog Number | Mfr. No. | Includes | Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Type | Quantity | Price | Quantity & Availability | |||
KL-K40000
![]() ![]() |
Koehler™ Instrument
K40000 |
0.25 in. Needle valves for purging and admitting LPG samples, dip tube with hook, knurled, threaded cap with O-ring gasket hand tightens to a positive seal | LPG Copper Strip Corrosion Test Cylinder | One unit per pack |
Each for $2,794.80
|
|
|||
KL-K40200
![]() ![]() |
Koehler™ Instrument
K40200 |
3.2mm Hole per ASTM specifications | Copper Strip | Each |
Each for $61.10
|
|
|||
Specifications
ASTM D1838, GPA 2140, ISO 6251, CE | |
Stainless Steel |