Learn More
Medchemexpress LLC Kojic acid | 501-30-4 | MFCD00006580 | 100.0% | 142.11 g/mol | C6H6O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0501545G
Encompass_Preferred
Kojic acid is a small-molecule tyrosinase inhibitor used in biochemical and cell-based research to reduce melanin synthesis. It chelates copper ions at the tyrosinase active site and shows antioxidant and antimicrobial activity.
  • Inhibits tyrosinase and reduces melanin production.
  • High purity suitable for biochemical assays.
  • Available as solid and DMSO solution formats, including 5 g solid.
  • Soluble in common laboratory solvents for reconstitution.
  • Useful in pigmentation, dermatology, and cell-based studies.
Catalog No. 50-002-09865