Learn More
Medchemexpress LLC Kojic acid | 501-30-4 | MFCD00006580 | 142.11 g/mol | C6H6O4 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW050154
Encompass_Preferred
Kojic acid is a small-molecule tyrosinase inhibitor supplied as a solid for research use in biochemical and cell-based studies of pigmentation, antioxidant activity, and related signaling pathways.
  • Tyrosinase inhibitor; inhibits melanin synthesis by chelating copper in the enzyme active site.
  • Solid form suitable for analytical and research applications.
  • Molecular formula C6H6O4; molecular weight 142.11 g/mol.
  • Common applications include pigmentation studies, antioxidant assays, and signaling pathway research.
Catalog No. 50-002-08853