Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kojic Acid Dipalmitate 98.0+%, TCI America™
SDP

Catalog No. K005025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K005025G 25 g
K00505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K005025G Supplier TCI America Supplier No. K005025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79725-98-7
Molecular Formula C38H66O6
MDL Number MFCD03840587
Synonym 5-(Palmitoyloxy)-2-[(palmitoyloxy)methyl]-4H-pyran-4-one

Specifications

Melting Point 96°C
Color White
Quantity 25 g
Formula Weight 618.94
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Kojic Acid Dipalmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.