Learn More
KIMBLE™ KONTES™ Allihn Condenser with Full Length Joints
SDP
Catalog No. 01672729 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No:
71/60
Catalog No. Standard Taper Joint No
01-672-729 71/60
1 options
Catalog No. 01-672-729 Supplier KIMBLE™ Supplier No. 4560000024
Allihn Condenser with water-cooled standard taper inner drip joint at bottom.
  • Straight outlet tube at top
  • Water-cooled Standard Taper inner drip joint at bottom
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Standard Taper Joint No 71/60
For Use With (Application) Refluxing Operations
For Use With (Equipment) Round-bottom Flask, Distillation Head
Length (Metric) Jacket 350 mm
Type Allihn Condenser
Material Borosilicate Glass