Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Packing Reservoir, with Joints
SDP
Catalog No. p-3246101 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
K4205700224 250 mL
K4205701024 1000 mL
K4205700524 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K4205700224 Supplier DWK Life Sciences Supplier No. 4205700224
Only null left
Add to Cart
Add to Cart
These heavy duty reservoirs are designed for use with 420510 and 420550 series chromatography columns with and without fritted glass supports or in combination with 420575 series flow control adapters.
  • Reservoirs are available in a variety of sizes and joint configurations
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Type Packing Reservoir with Joints
Volume (Metric) 250 mL