Learn More
DWK Life Sciences Kimble™ Kontes™ Connecting Distillation Adapter with Parallel 14/20 Standard Taper Joints
SDP
Catalog No. K2750501420 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
14/20
Catalog No. Standard Taper Joint No.
K275050-1420 14/20
1 options
Catalog No. K275050-1420 Supplier DWK Life Sciences Supplier No. 2750501420
Add to Cart
Add to Cart
Useful in the sub-boiling separation of mixtures having volatile components.
  • Standard Taper inner joints are parallel
  • Joints are the same size, 100mm between joints
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) Useful in the sub-boiling separation of mixtures having volatile components
Material Borosilicate Glass
Product Type Connecting Distillation Adapter
Standard Taper Joint No. 14/20