Learn More
DWK Life Sciences Kimble™ Liebig Medium Length Condenser with 14/20 Joints
SDP
Catalog No. K2822100000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No:
14/20
Catalog No. Standard Taper Joint No
K282210-0000 14/20
1 options
Catalog No. K282210-0000 Supplier DWK Life Sciences Supplier No. 2822100000
Add to Cart
Add to Cart
Design eliminates the use of bulbs where holdup is likely to occur.
  • Lower inner drip joint is designed to extend into 287800-0000 distillation receiver
  • Approximate Condensing Area 34cm2
  • Overall height 178mm
  • Fits tubing ID 1/4 in.
  • Standard Taper joints 14/20
  • Design conforms to NSN 6640-01-346-9222
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Standard Taper Joint No 14/20
Height (Metric) 178 mm
Height (English) 7 in.
Type Condenser
For Use With (Application) Lower inner drip joint is designed to extend into 287800-0000 distillation receiver
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438, Type I, Class A requirements