Description
- Ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers
- Support base has a coarse porosity glass frit and an integral vacuum connection, located above the drip tip that prevents contamination of the vacuum line with filtrate droplets
Specifications
Specifications
| For Use With (Equipment) | Microfiltration Apparatus |
| Material | Glass |
| Type | Funnel (500 mL), Flask (2 L) |
| Volume (Metric) | 500 mL, 2 L |