Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ NMR Polyethylene Sample Tube Pressure Caps for 5 and 10mm Tubes
SDP

Catalog No. K8970950021 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Assorted
Black
Green
Red
White
Yellow
For Use With (Equipment):
10 mm NMR Sample Tube
3 mm NMR Sample Tube
5 mm NMR Sample Tube
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color For Use With (Equipment)
K897095-0021 Black 5 mm NMR Sample Tube
K897095-0081 Yellow 5 mm NMR Sample Tube
K897095-0061 White 5 mm NMR Sample Tube
K897095-1001 Red 10 mm NMR Sample Tube
K897095-1021 Black 10 mm NMR Sample Tube
K8970930001 Red 3 mm NMR Sample Tube
K897095-0001 Red 5 mm NMR Sample Tube
K897095-0012 Assorted 5 mm NMR Sample Tube
K897095-0071 Green 5 mm NMR Sample Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. K897095-0021 Supplier DWK Life Sciences Supplier No. 8970950021
Add to Cart
Add to Cart

Tight-fitting, polyethylene cap which stays in place even with a build-up of internal vaporization pressures

TRUSTED_SUSTAINABILITY

Specifications

Product Type Pressure Cap
Material Polyethylene
Color Black
For Use With (Application) Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures
For Use With (Equipment) 5 mm NMR Sample Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.