Learn More
DWK LIFE SCIENCES™ KIMBLE™ KONTES™ NMR Single and Double Sealing Manifold
Shop All DWK Life Sciences Products

Description
- Enables easy evacuation and tip-off of NMR tubes without additional sealing to other glass apparatus
- Available in single and double port configurations
- Equipped with KONTES™ HI-VAC™ valves for individual port control
- Compatible with NMR tubes from leading manufacturers including Bruker, Jeol, and Agilent
Specifications
Specifications
| Product Type | Single NMR Sealing Manifold |
| Autoclavable | Autoclavable |
| Material | Borosilicate Glass |
| Color | Translucent |
| Sterility | Non-sterile |
| For Use With (Equipment) | Complete NMR Tube Sealing Manifold |
| Disposable | Reusable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.