Description
Outer joint at top; inner joint at bottom
Specifications
Specifications
| Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
| For Use With (Application) | For connecting dissimilar joints |
| Material | Borosilicate Glass |
| Product Type | Reducing Adapter |
| Standard Taper Joint No. | 45/50 Outer Top, 24/40 Inner Bottom |