Learn More
DWK Life Sciences Kimble™ Kontes™ Reducing Adapters
SDP
Catalog No. K2747500263 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No.:
19/22 Outer Top, 14/20 Inner Bottom
24/40 Outer Top, 14/20 Inner Bottom
24/40 Outer Top, 19/22 Inner Bottom
29/42 Outer Top, 24/40 Inner Bottom
45/50 Outer Top, 24/40 Inner Bottom
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
K274750-0263 24/40 Outer Top, 14/20 Inner Bottom
K150100-0274 29/42 Outer Top, 24/40 Inner Bottom
K150100-0432 45/50 Outer Top, 24/40 Inner Bottom
K274750-0253 19/22 Outer Top, 14/20 Inner Bottom
K274750-0264 24/40 Outer Top, 19/22 Inner Bottom
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. K274750-0263 Supplier DWK Life Sciences Supplier No. 2747500263
For connecting dissimilar standard taper joints
Outer joint at top; inner joint at bottom
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Application) For connecting dissimilar joints
Material Borosilicate Glass
Product Type Reducing Adapter
Standard Taper Joint No. 24/40 Outer Top, 14/20 Inner Bottom