Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Medium Length Stoppers
Shop All DWK Life Sciences Products
Description
- All stoppers are solid except Standard Taper stopper sizes 32 and 38, which are hollow
- Made in accordance with ASTM Standard E694
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| For Use With (Equipment) | Flasks, mixing cylinders and separatory funnels |
| Material | Glass |
| Product Type | Solid Pennyhead Stopper |