Learn More
DWK Life Sciences Kimble™ Kontes™ Solid Pennyhead Glass Medium Length Stoppers
GSA_VASDP
Catalog No. K8501000022 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
10/18
19
19/22
24/25
27
38
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
K850100-0022 -
K850100-0019 19
K850500-1018 10/18
K850100-0027 27
02-543-27 38
K850500-2425 24/25
K850500-1922 19/22
22-301346 -
22-301347 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. K850100-0022 Supplier DWK Life Sciences Supplier No. 8501000022
Add to Cart
Add to Cart
Stoppers are used with laboratory glassware such as flasks, mixing cylinders and separatory funnels.
  • All stoppers are solid except Standard Taper stopper sizes 32 and 38, which are hollow
  • Made in accordance with ASTM Standard E694
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Product Type Solid Pennyhead Stopper
For Use With (Equipment) Flasks, mixing cylinders and separatory funnels