Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column PEEK Nut for 3/16 in. Tubing
SDP
Catalog No. K4208213519 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
For 0.18 in. O.D. Tubing
Catalog No. For Use With (Equipment)
K420821-3519 For 0.18 in. O.D. Tubing
1 options
Catalog No. K420821-3519 Supplier DWK Life Sciences Supplier No. 4208213519
Add to Cart
Add to Cart
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
  • PEEK Nut for 3/16 in. OD Tubing
  • 5/16 in.-24 Thread
  • Use with the 420822-Series Flangeless Ferrules
  • Case of 1
  • Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
TRUSTED_SUSTAINABILITY
Specifications
Material PEEK
For Use With (Equipment) For 0.18 in. O.D. Tubing
Type Tubing Nut
Thread Size 0.312 to 24 UNF
Quantity Each