Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column EFTE Ferrule for 3/16 in. Tubing
SDP
Catalog No. K4208221019 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
For 0.18 in. O.D. Tubing
Catalog No. For Use With (Equipment)
K420822-1019 For 0.18 in. O.D. Tubing
1 options
Catalog No. K420822-1019 Supplier DWK Life Sciences Supplier No. 4208221019
Add to Cart
Add to Cart
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
  • EFTE Flangeless Ferrule for 3/16 in. OD Tubing
  • For use with 420821 Series Male Nuts
  • Case of 1
  • Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
TRUSTED_SUSTAINABILITY
Specifications
Material ETFE
For Use With (Equipment) For 0.18 in. O.D. Tubing
Type Ferrule
Quantity Each